Product Details

SNP ID
rs189308436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:31002177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCAGCTTCCGACACAATGTAA[A/G]GAATATCTGTGCTATGAGAGCGCGT
Phenotype
MIM: 603576
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRPM1 PubMed Links

Gene Details

Gene
TRPM1
Gene Name
transient receptor potential cation channel subfamily M member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252020.1 4888 Missense Mutation NP_001238949.1
NM_001252024.1 4888 Missense Mutation NP_001238953.1
NM_001252030.1 4888 Intron NP_001238959.1
NM_002420.5 4888 Missense Mutation NP_002411.3

View Full Product Details