Product Details

SNP ID
rs189343001
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:158420447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGCAGCCACTAGTCCCATACA[C/T]ACCCCATGTCCCATTAGCACTGAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10K2 PubMed Links

Gene Details

Gene
OR10K2
Gene Name
olfactory receptor family 10 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004476.1 420 Silent Mutation GTA,GTG V140V NP_001004476.1

View Full Product Details