Product Details

SNP ID
rs189370242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218990981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCCCTCAAACAGTGTCACACGG[A/C]TGTCATTGTGGTTCTGAGGCACAGC
Phenotype
MIM: 600836 MIM: 607150
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRYBA2 PubMed Links

Gene Details

Gene
CRYBA2
Gene Name
crystallin beta A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057093.1 352 Missense Mutation AGC,ATC S106I NP_476434.1
NM_057094.1 352 Missense Mutation AGC,ATC S106I NP_476435.1
Gene
FEV
Gene Name
FEV, ETS transcription factor
There are no transcripts associated with this gene.

View Full Product Details