Product Details

SNP ID
rs189183340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151176570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCTTGGAATCTGACCTGCAGA[A/G]TTTCAGGAGTCACCAGGCCGACGGC
Phenotype
MIM: 615054 MIM: 603354
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB10 PubMed Links

Gene Details

Gene
ASB10
Gene Name
ankyrin repeat and SOCS box containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142459.1 1472 Missense Mutation ACT,ATT T404I NP_001135931.2
NM_001142460.1 1472 Intron NP_001135932.2
NM_080871.3 1472 Missense Mutation ACT,ATT T389I NP_543147.2
Gene
GBX1
Gene Name
gastrulation brain homeobox 1
There are no transcripts associated with this gene.

View Full Product Details