Product Details

SNP ID
rs189570280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:26561154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTGGCATGGCTGAATCATTCA[A/G]AGTTGGATTGTAGTAGCTAGAATTC
Phenotype
MIM: 610110 MIM: 608566
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANO3 PubMed Links

Gene Details

Gene
ANO3
Gene Name
anoctamin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313726.1 1270 Intron NP_001300655.1
NM_001313727.1 1270 Intron NP_001300656.1
NM_031418.3 1270 Intron NP_113606.2
XM_011520282.2 1270 Intron XP_011518584.1
XM_017018118.1 1270 Intron XP_016873607.1
XM_017018119.1 1270 Intron XP_016873608.1
Gene
MUC15
Gene Name
mucin 15, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135091.1 1270 Silent Mutation CTG,TTG L333L NP_001128563.1
NM_001135092.1 1270 Silent Mutation CTG,TTG L283L NP_001128564.1
NM_145650.3 1270 Silent Mutation CTG,TTG L306L NP_663625.2
XM_017017235.1 1270 Silent Mutation CTG,TTG L422L XP_016872724.1
XM_017017236.1 1270 Silent Mutation CTG,TTG L372L XP_016872725.1

View Full Product Details