Product Details

SNP ID
rs189570291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:18795525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCACCAACCCACAGCTCTGCGT[C/T]GAGTTTTTACTAGATTGCTGCAAAT
Phenotype
MIM: 607669 MIM: 603674 MIM: 607032
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL6IP1 PubMed Links

Gene Details

Gene
ARL6IP1
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313858.1 559 Missense Mutation CAA,CGA Q87R NP_001300787.1
NM_015161.2 559 Missense Mutation CAA,CGA Q116R NP_055976.1
Gene
RPS15A
Gene Name
ribosomal protein S15a
There are no transcripts associated with this gene.

Gene
SMG1
Gene Name
SMG1, nonsense mediated mRNA decay associated PI3K related kinase
There are no transcripts associated with this gene.

View Full Product Details