Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001178030.1 | 2034 | Intron | NP_001171501.1 | ||
NM_001178031.1 | 2034 | Silent Mutation | TTC,TTT | F600F | NP_001171502.1 |
NM_004630.3 | 2034 | Silent Mutation | TTC,TTT | F626F | NP_004621.2 |
NM_201995.2 | 2034 | Intron | NP_973724.1 | ||
NM_201997.2 | 2034 | Intron | NP_973726.2 | ||
NM_201998.2 | 2034 | Intron | NP_973727.1 | ||
XM_011545244.1 | 2034 | Intron | XP_011543546.1 | ||
XM_011545245.1 | 2034 | UTR 3 | XP_011543547.1 | ||
XM_011545246.1 | 2034 | UTR 3 | XP_011543548.1 | ||
XM_011545247.1 | 2034 | Intron | XP_011543549.1 | ||
XM_011545248.1 | 2034 | Intron | XP_011543550.1 | ||
XM_017018244.1 | 2034 | Silent Mutation | TTC,TTT | F600F | XP_016873733.1 |
XM_017018245.1 | 2034 | Silent Mutation | TTC,TTT | F630F | XP_016873734.1 |
XM_017018246.1 | 2034 | Intron | XP_016873735.1 | ||
XM_017018247.1 | 2034 | UTR 3 | XP_016873736.1 | ||
XM_017018248.1 | 2034 | Intron | XP_016873737.1 | ||
XM_017018249.1 | 2034 | Silent Mutation | TTC,TTT | F511F | XP_016873738.1 |
XM_017018250.1 | 2034 | Silent Mutation | TTC,TTT | F511F | XP_016873739.1 |
XM_017018251.1 | 2034 | Silent Mutation | TTC,TTT | F511F | XP_016873740.1 |