Product Details

SNP ID
rs189944635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:85107333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAATAAAAATAGGCTTACTTTAA[A/T]CAGTCGCAAAATTTCTGGGCAGTCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SATL1 PubMed Links

Gene Details

Gene
SATL1
Gene Name
spermidine/spermine N1-acetyl transferase-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012980.2 1636 Missense Mutation ATT,TTT I546F NP_001012998.2
XM_011530940.2 1636 Missense Mutation ATT,TTT I546F XP_011529242.1

View Full Product Details