Product Details

SNP ID
rs189878543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194641210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTACATACCATACAATTCACCAA[C/T]GTAAAGTGTGTAATTCAATGGTCTT
Phenotype
MIM: 610780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LSG1 PubMed Links

Gene Details

Gene
LSG1
Gene Name
large 60S subunit nuclear export GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018385.2 2478 UTR 3 NP_060855.2
XM_017006796.1 2478 UTR 3 XP_016862285.1
Gene
TMEM44
Gene Name
transmembrane protein 44
There are no transcripts associated with this gene.

View Full Product Details