Product Details

SNP ID
rs189583607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12043814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATCCATAGAGTTTCTAGCCAGC[A/G]TGAGTCCTTACATGCTTTTGAAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928464 PubMed Links

Gene Details

Gene
LOC101928464
Gene Name
uncharacterized LOC101928464
There are no transcripts associated with this gene.

Gene
ZNF433
Gene Name
zinc finger protein 433
There are no transcripts associated with this gene.

Gene
ZNF878
Gene Name
zinc finger protein 878
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080404.2 1753 Silent Mutation CAC,CAT H529H NP_001073873.2
XM_017027195.1 1753 Intron XP_016882684.1
XM_017027196.1 1753 Intron XP_016882685.1

View Full Product Details