Product Details

SNP ID
rs189650562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17271653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCCAAAGCTGGAGTCGTTCAA[C/T]GGGTAAGAGGGACATTTTAGGGCTT
Phenotype
MIM: 612766 MIM: 611810
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKLE1 PubMed Links

Gene Details

Gene
ANKLE1
Gene Name
ankyrin repeat and LEM domain containing 1
There are no transcripts associated with this gene.

Gene
BABAM1
Gene Name
BRISC and BRCA1 A complex member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033549.2 545 Silent Mutation AAC,AAT N114N NP_001028721.1
NM_001288756.1 545 Silent Mutation AAC,AAT N114N NP_001275685.1
NM_001288757.1 545 Silent Mutation AAC,AAT N114N NP_001275686.1
NM_014173.3 545 Silent Mutation AAC,AAT N114N NP_054892.2
Gene
USHBP1
Gene Name
USH1 protein network component harmonin binding protein 1
There are no transcripts associated with this gene.

View Full Product Details