Product Details

SNP ID
rs189744812
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:85159154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCAGTACCACTCAAATTTAACA[C/T]ATGAGGTCGTTCTTTCTTCTGCCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SYDE2 PubMed Links

Gene Details

Gene
SYDE2
Gene Name
synapse defective Rho GTPase homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032184.1 2617 Missense Mutation ATG,GTG M1061V NP_115560.1
XM_005271254.3 2617 Intron XP_005271311.1
XM_006710946.3 2617 Intron XP_006711009.1
XM_017002483.1 2617 Missense Mutation TAT,TGT Y1256C XP_016857972.1
XM_017002484.1 2617 Intron XP_016857973.1
XM_017002485.1 2617 Missense Mutation ATG,GTG M803V XP_016857974.1
XM_017002486.1 2617 Missense Mutation ATG,GTG M803V XP_016857975.1

View Full Product Details