Product Details

SNP ID
rs189448963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:104381289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGACTTCTCCCTTATAGGGTGCA[C/T]GCTCCACAGGGGTAGGTGTGCAAGT
Phenotype
MIM: 613326
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPYS PubMed Links

Gene Details

Gene
DPYS
Gene Name
dihydropyrimidinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001385.2 1572 Missense Mutation CAT,CGT H490R NP_001376.1
XM_005250818.3 1572 Missense Mutation CAT,CGT H526R XP_005250875.1
XM_006716518.3 1572 Missense Mutation CAT,CGT H473R XP_006716581.1
XM_011516903.2 1572 Intron XP_011515205.1
XM_017013167.1 1572 Intron XP_016868656.1

View Full Product Details