Product Details

SNP ID
rs189477293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7620353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCGTGTTGTACGTGGCCTGCA[C/T]GCTGCGCCGGATACGCAGCATTTCC
Phenotype
MIM: 612685 MIM: 614770 MIM: 610850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAMSAP3 PubMed Links

Gene Details

Gene
CAMSAP3
Gene Name
calmodulin regulated spectrin associated protein family member 3
There are no transcripts associated with this gene.

Gene
MIR6792
Gene Name
microRNA 6792
There are no transcripts associated with this gene.

Gene
PET100
Gene Name
PET100 homolog
There are no transcripts associated with this gene.

Gene
XAB2
Gene Name
XPA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020196.2 2214 Missense Mutation ATG,GTG M730V NP_064581.2

View Full Product Details