Product Details

SNP ID
rs190336639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81913784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGGCTCACTCACCTGCTATAGG[C/T]GGGGATCTCCAAGCCCAGCTCGGCC
Phenotype
MIM: 602020 MIM: 602679 MIM: 606212
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAFG PubMed Links

Gene Details

Gene
MAFG
Gene Name
MAF bZIP transcription factor G
There are no transcripts associated with this gene.

Gene
PCYT2
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
There are no transcripts associated with this gene.

Gene
SIRT7
Gene Name
sirtuin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016538.2 2648 Missense Mutation ACC,GCC T332A NP_057622.1
XM_006722284.2 2648 Missense Mutation ACC,GCC T250A XP_006722347.1
XM_011523580.1 2648 Missense Mutation ACC,GCC T297A XP_011521882.1
XM_011523581.2 2648 Missense Mutation ACC,GCC T261A XP_011521883.1

View Full Product Details