Product Details

SNP ID
rs190500642
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:32889093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATGGTGACGCCTCCAAGTTTGC[C/T]GAGCACGTCTTCCGCACCTTTGACA
Phenotype
MIM: 142622
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HPCA PubMed Links

Gene Details

Gene
HPCA
Gene Name
hippocalcin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002143.2 707 Silent Mutation GCC,GCT A65A NP_002134.2
XM_005270792.2 707 Silent Mutation GCC,GCT A65A XP_005270849.1
XM_017001118.1 707 Silent Mutation GCC,GCT A65A XP_016856607.1
Gene
TMEM54
Gene Name
transmembrane protein 54
There are no transcripts associated with this gene.

View Full Product Details