Product Details

SNP ID
rs190463100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:37934433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATACAGCTGAAGTTAATCTGAC[C/T]GTTCTTGGGGATATAGATGTGCTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOT1L1 PubMed Links

Gene Details

Gene
GOT1L1
Gene Name
glutamic-oxaloacetic transaminase 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152413.2 1506 Missense Mutation AGT,GGT S376G NP_689626.2
XM_005273399.3 1506 Missense Mutation AGT,GGT S375G XP_005273456.1
XM_006716285.3 1506 Missense Mutation AGT,GGT S345G XP_006716348.1
XM_011544391.2 1506 Missense Mutation AGT,GGT S194G XP_011542693.1

View Full Product Details