Product Details

SNP ID
rs190437417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7565023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCCTCAGCACTGATGAGGTAGT[A/G]GAGAAGCTGGAGGACATTTTCCAGC
Phenotype
MIM: 602641 MIM: 612844 MIM: 602695 MIM: 604472
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF4A1 PubMed Links

Gene Details

Gene
EIF4A1
Gene Name
eukaryotic translation initiation factor 4A1
There are no transcripts associated with this gene.

Gene
SENP3
Gene Name
SUMO1/sentrin/SMT3 specific peptidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015670.5 1303 Silent Mutation GTA,GTG V340V NP_056485.2
Gene
SENP3-EIF4A1
Gene Name
SENP3-EIF4A1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
SNORA48
Gene Name
small nucleolar RNA, H/ACA box 48
There are no transcripts associated with this gene.

Gene
TNFSF12
Gene Name
tumor necrosis factor superfamily member 12
There are no transcripts associated with this gene.

Gene
TNFSF12-TNFSF13
Gene Name
TNFSF12-TNFSF13 readthrough
There are no transcripts associated with this gene.

Gene
TNFSF13
Gene Name
tumor necrosis factor superfamily member 13
There are no transcripts associated with this gene.

View Full Product Details