Product Details

SNP ID
rs190126592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48352107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTGAAAGATACTCACCTTCGGG[A/T]AGATTCCCTGGAGCCTGCCGAAAGT
Phenotype
MIM: 300325
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SSX3 PubMed Links

Gene Details

Gene
SSX3
Gene Name
SSX family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021014.3 Intron NP_066294.1
XM_011543885.2 Intron XP_011542187.1

View Full Product Details