Product Details

SNP ID
rs190226328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46124648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAAGATGCAGCATCGTGGCCTC[A/T]TGCTTCCAAAGATGCTCTAGGAGAA
Phenotype
MIM: 610545 MIM: 610546
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IGFL2 PubMed Links

Gene Details

Gene
IGFL2
Gene Name
IGF like family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002915.2 2454 Intron NP_001002915.2
NM_001135113.1 2454 Intron NP_001128585.1
XM_011526501.1 2454 Intron XP_011524803.1
XM_017026335.1 2454 Intron XP_016881824.1
Gene
IGFL3
Gene Name
IGF like family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207393.1 2454 Missense Mutation AAG,ATG K1M NP_997276.1
XM_011526953.2 2454 Missense Mutation AAG,ATG K1M XP_011525255.1
XM_017026796.1 2454 Missense Mutation AAG,ATG K1M XP_016882285.1
XM_017026797.1 2454 Missense Mutation AAG,ATG K1M XP_016882286.1

View Full Product Details