Product Details
- SNP ID
-
rs190269020
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:200871444 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATAGTTATGTCCTTAATGTGTACA[G/T]CATTAAGAGGTCGGTATGTCTTCTC
- Phenotype
-
MIM: 601951
MIM: 615811
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CLK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7562391] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLK1
- Gene Name
- CDC like kinase 1
There are no transcripts associated with this gene.
- Gene
- PPIL3
- Gene Name
- peptidylprolyl isomerase like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032472.3 |
700 |
Missense Mutation |
GAT,GCT |
D150A |
NP_115861.1 |
NM_130906.2 |
700 |
Missense Mutation |
GAT,GCT |
D146A |
NP_570981.1 |
XM_005246651.3 |
700 |
Missense Mutation |
GAT,GCT |
D150A |
XP_005246708.1 |
XM_005246652.4 |
700 |
Missense Mutation |
GAT,GCT |
D146A |
XP_005246709.1 |
XM_011511358.2 |
700 |
Missense Mutation |
GAT,GCT |
D146A |
XP_011509660.1 |
XM_017004352.1 |
700 |
Missense Mutation |
GAT,GCT |
D150A |
XP_016859841.1 |
XM_017004353.1 |
700 |
Missense Mutation |
GAT,GCT |
D150A |
XP_016859842.1 |
XM_017004354.1 |
700 |
Intron |
|
|
XP_016859843.1 |
XM_017004355.1 |
700 |
Intron |
|
|
XP_016859844.1 |
XM_017004356.1 |
700 |
Intron |
|
|
XP_016859845.1 |
XM_017004357.1 |
700 |
Missense Mutation |
GAT,GCT |
D97A |
XP_016859846.1 |
XM_017004358.1 |
700 |
Missense Mutation |
GAT,GCT |
D97A |
XP_016859847.1 |
XM_017004359.1 |
700 |
Missense Mutation |
GAT,GCT |
D97A |
XP_016859848.1 |
XM_017004360.1 |
700 |
Missense Mutation |
GAT,GCT |
D97A |
XP_016859849.1 |
View Full Product Details