Product Details

SNP ID
rs190269143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108623246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTCGGTGGGGGAGTAATTACG[A/C]ACGGGGTACATGTGGCCCTTGCGGG
Phenotype
MIM: 600738
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SELPLG PubMed Links

Gene Details

Gene
SELPLG
Gene Name
selectin P ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206609.1 1287 Silent Mutation GTG,GTT V370V NP_001193538.1
NM_003006.4 1287 Silent Mutation GTG,GTT V354V NP_002997.2

View Full Product Details