Product Details

SNP ID
rs190024469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50872273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGTGGGACATGGTCGACCGCAA[C/T]CTGCGGTGAGTACCGCCCGGTCCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD12B PubMed Links

Gene Details

Gene
ABHD12B
Gene Name
abhydrolase domain containing 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206673.1 114 Silent Mutation AAC,AAT N33N NP_001193602.1
NM_181533.3 114 UTR 5 NP_853511.2
NM_181814.1 114 Silent Mutation AAC,AAT N33N NP_861535.1
XM_006720052.1 114 Silent Mutation AAC,AAT N33N XP_006720115.1
XM_011536472.1 114 Silent Mutation AAC,AAT N33N XP_011534774.1
XM_011536474.2 114 Intron XP_011534776.1
XM_017021031.1 114 UTR 5 XP_016876520.1

View Full Product Details