Product Details

SNP ID
rs190637588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:129376185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCTCTACTCCGTGTGCCCGTC[A/G]CTGGTCGTGCGACCCAAGCCCCTGC
Phenotype
MIM: 604823
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BARX2 PubMed Links

Gene Details

Gene
BARX2
Gene Name
BARX homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003658.4 200 Silent Mutation TCA,TCG S50S NP_003649.2
XM_011543043.1 200 Intron XP_011541345.1
XM_011543044.2 200 Intron XP_011541346.1

View Full Product Details