Product Details

SNP ID
rs190657184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197572124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGCTGTTGTGCTGTGCCAGCC[A/T]TCCCTCCTTTGTCCAGCATTCCCGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RFTN2 PubMed Links

Gene Details

Gene
RFTN2
Gene Name
raftlin family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144629.2 2009 Missense Mutation AGG,TGG R464W NP_653230.2
XM_011510595.2 2009 Missense Mutation AGG,TGG R474W XP_011508897.1
XM_011510596.2 2009 Missense Mutation AGG,TGG R473W XP_011508898.1
XM_011510597.2 2009 Missense Mutation AGG,TGG R463W XP_011508899.1
XM_011510598.2 2009 Missense Mutation AGG,TGG R457W XP_011508900.1
XM_017003332.1 2009 Missense Mutation AGG,TGG R447W XP_016858821.1

View Full Product Details