Product Details
- SNP ID
-
hCV186185060
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:184230801 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGTGGTGTCCGGCTTCGAGGCCG[A/G]GGCCGCCGGACGGCGCGTCTCCTTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VWA5B2
PubMed Links
Gene Details
- Gene
- VWA5B2
- Gene Name
- von Willebrand factor A domain containing 5B2
There are no transcripts associated with this gene.
View Full Product Details