Product Details

SNP ID
rs190754463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27094495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGACCCCGGGGAAGGAACGCAG[A/G]GCGAAGAGCTGGACTTCTCTGAGGA
Phenotype
MIM: 142955 MIM: 604685
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOTAIRM1 PubMed Links

Gene Details

Gene
HOTAIRM1
Gene Name
HOXA transcript antisense RNA, myeloid-specific 1
There are no transcripts associated with this gene.

Gene
HOXA1
Gene Name
homeobox A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005522.4 844 Missense Mutation CCC,CTC P318L NP_005513.1
NM_153620.2 844 UTR 3 NP_705873.2
Gene
HOXA2
Gene Name
homeobox A2
There are no transcripts associated with this gene.

View Full Product Details