Product Details

SNP ID
rs190764556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61509200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGAGGAGATCGAGGAGCTGCGC[C/G]AGCTGCGCATCCTGGCGCTGGACTT
Phenotype
MIM: 604146
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRRC10B PubMed Links

Gene Details

Gene
LRRC10B
Gene Name
leucine rich repeat containing 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145077.1 401 Missense Mutation CAG,GAG Q68E NP_001138549.1
Gene
MIR4488
Gene Name
microRNA 4488
There are no transcripts associated with this gene.

Gene
SYT7
Gene Name
synaptotagmin 7
There are no transcripts associated with this gene.

View Full Product Details