Product Details

SNP ID
rs190827633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67280960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGCTTCAGTCTGAGTGCAGCA[A/G]CCAGGAACTCATCCGCCTCCTGCTG
Phenotype
MIM: 609526 MIM: 600477
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD19 PubMed Links

Gene Details

Gene
KCTD19
Gene Name
potassium channel tetramerization domain containing 19
There are no transcripts associated with this gene.

Gene
PLEKHG4
Gene Name
pleckstrin homology and RhoGEF domain containing G4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129727.2 1019 Missense Mutation AAC,AGC N225S NP_001123199.1
NM_001129728.1 1019 Missense Mutation AAC,AGC N225S NP_001123200.1
NM_001129729.2 1019 Missense Mutation AAC,AGC N225S NP_001123201.1
NM_001129731.2 1019 Missense Mutation AAC,AGC N144S NP_001123203.1
XM_011522985.2 1019 Missense Mutation AAC,AGC N225S XP_011521287.1
XM_011522986.2 1019 Missense Mutation AAC,AGC N225S XP_011521288.1
XM_011522987.2 1019 Missense Mutation AAC,AGC N225S XP_011521289.1
XM_011522988.2 1019 Missense Mutation AAC,AGC N225S XP_011521290.1
Gene
SLC9A5
Gene Name
solute carrier family 9 member A5
There are no transcripts associated with this gene.

View Full Product Details