Product Details

SNP ID
rs190834047
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:241660600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTCCAGATGTGAAGAATCAAGA[C/T]GTGAAACGCTTTTACAGGAAACTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR64 PubMed Links

Gene Details

Gene
WDR64
Gene Name
WD repeat domain 64
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144625.4 423 Silent Mutation GAC,GAT D72D NP_653226.4
XM_006711736.3 423 Silent Mutation GAC,GAT D72D XP_006711799.1
XM_011544085.2 423 Intron XP_011542387.1
XM_011544086.2 423 Intron XP_011542388.1
XM_011544087.2 423 Silent Mutation GAC,GAT D72D XP_011542389.1
XM_011544091.1 423 Silent Mutation GAC,GAT D72D XP_011542393.1
XM_011544092.2 423 Silent Mutation GAC,GAT D72D XP_011542394.1
XM_017000315.1 423 UTR 5 XP_016855804.1

View Full Product Details