Product Details

SNP ID
rs191177224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41958740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGCCCGGGAGGCACTCACACAC[A/T]GTACAGGATGAGGCCCAGGAACACG
Phenotype
MIM: 182350 MIM: 604925
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP1A3 PubMed Links

Gene Details

Gene
ATP1A3
Gene Name
ATPase Na+/K+ transporting subunit alpha 3
There are no transcripts associated with this gene.

Gene
RABAC1
Gene Name
Rab acceptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006423.2 349 Missense Mutation AGT,TGT S89C NP_006414.2

View Full Product Details