Product Details
- SNP ID
-
rs191186879
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:28593320 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCTCAGCTCCCACCACTCTTGCA[C/T]GTGCTGGTACCAGGACCCATAGGAC
- Phenotype
-
MIM: 613374
MIM: 601292
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SGF29
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3743963] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SGF29
- Gene Name
- SAGA complex associated factor 29
There are no transcripts associated with this gene.
- Gene
- SULT1A2
- Gene Name
- sulfotransferase family 1A member 2
View Full Product Details