Product Details

SNP ID
rs191443110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219172586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAATGCATTCCTCACAGCCCTAC[A/C]TAGCCTGGGAAAACGAAAGACTTGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CNPPD1 PubMed Links

Gene Details

Gene
CNPPD1
Gene Name
cyclin Pas1/PHO80 domain containing 1
There are no transcripts associated with this gene.

Gene
FAM134A
Gene Name
family with sequence similarity 134 member A
There are no transcripts associated with this gene.

Gene
SLC23A3
Gene Name
solute carrier family 23 member 3
There are no transcripts associated with this gene.

View Full Product Details