Product Details

SNP ID
rs191448565
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.6:32554876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTCTGGAACCACCTGACTTTAA[C/T]GCTGCCTGGATAGAAACCACTCACA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HLA-DRB6 PubMed Links

Gene Details

Gene
HLA-DRB6
Gene Name
major histocompatibility complex, class II, DR beta 6 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details