Product Details

SNP ID
rs191600199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112255438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTTCAGGAGCTTGCCACTGTGC[C/T]TCTAAGACCGTCATGTATTGATCTT
Phenotype
MIM: 611904
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100132686 PubMed Links

Gene Details

Gene
LOC100132686
Gene Name
uncharacterized LOC100132686
There are no transcripts associated with this gene.

Gene
PLET1
Gene Name
placenta expressed transcript 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145024.1 607 Silent Mutation GAA,GAG E112E NP_001138496.1

View Full Product Details