Product Details

SNP ID
rs191570000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123775169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTAAACATGTCAAGAAAGATGA[A/G]CGCAGATACTATGAGGAACTGCTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM91A1 PubMed Links

Gene Details

Gene
FAM91A1
Gene Name
family with sequence similarity 91 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317917.1 444 UTR 5 NP_001304846.1
NM_001317918.1 444 Silent Mutation GAA,GAG E60E NP_001304847.1
NM_144963.3 444 Silent Mutation GAA,GAG E60E NP_659400.2

View Full Product Details