Product Details

SNP ID
rs191140439
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:1460829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGCATCGACGTGATGGTCCAC[C/T]TCGACCTGCCAGGGCAGGAGGAGCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATAD3C PubMed Links

Gene Details

Gene
ATAD3C
Gene Name
ATPase family, AAA domain containing 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039211.2 1887 Missense Mutation CTC,TTC L298F NP_001034300.2

View Full Product Details