Product Details

SNP ID
rs191086352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64465539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCAAATATCCTTGAAAAACAAG[A/C]AAGTAAGAGAACTTTGTTGCGTGTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MILR1 PubMed Links

Gene Details

Gene
MILR1
Gene Name
mast cell immunoglobulin like receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085423.1 905 Missense Mutation GAA,GCA E284A NP_001078892.1
NM_001291316.1 905 Missense Mutation GAA,GCA E189A NP_001278245.1
NM_001291317.1 905 Missense Mutation GAA,GCA E194A NP_001278246.1
XM_017024486.1 905 Missense Mutation GAA,GCA E326A XP_016879975.1
XM_017024487.1 905 Missense Mutation GAA,GCA E226A XP_016879976.1

View Full Product Details