Product Details

SNP ID
rs191105016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81682108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGAGACCTAAGAGCTGCACACA[C/G]GATGCAGAGAGCTGGGTGGGGTCAG
Phenotype
MIM: 614271 MIM: 604375
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL16 PubMed Links

Gene Details

Gene
ARL16
Gene Name
ADP ribosylation factor like GTPase 16
There are no transcripts associated with this gene.

Gene
CCDC137
Gene Name
coiled-coil domain containing 137
There are no transcripts associated with this gene.

Gene
HGS
Gene Name
hepatocyte growth factor-regulated tyrosine kinase substrate
There are no transcripts associated with this gene.

View Full Product Details