Product Details
- SNP ID
-
rs191144864
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:224190018 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCTCTTTTATGCCTTTCGACCTC[A/T]GTTCATCAACCCCAAACCAATTACG
- Phenotype
-
MIM: 615843
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DEGS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6686449] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DEGS1
- Gene Name
- delta 4-desaturase, sphingolipid 1
View Full Product Details