Product Details

SNP ID
rs191719233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39344002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCACGTGCACAGCAGCATGGC[A/G]GCAGTGGCACCTGCTCCCGTGCAGT
Phenotype
MIM: 606710
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LHFP PubMed Links

Gene Details

Gene
LHFP
Gene Name
lipoma HMGIC fusion partner
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005780.2 1048 Silent Mutation GCC,GCT A179A NP_005771.1
XM_011534861.1 1048 Silent Mutation GCC,GCT A179A XP_011533163.1

View Full Product Details