Product Details

SNP ID
rs192149727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:151282275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTACCAAGATACAACGGCAGAC[A/G]TCGCCTTGTTGAAACTGTCCTCTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS48 PubMed Links

Gene Details

Gene
PRSS48
Gene Name
protease, serine 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183375.2 789 Missense Mutation ATC,GTC I115V NP_899231.2
XM_011531921.2 789 Missense Mutation ATC,GTC I138V XP_011530223.1
XM_011531922.2 789 Intron XP_011530224.1

View Full Product Details