Product Details
- SNP ID
-
rs192093174
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:46733727 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTAGGGAGGCAGACAGGCTGGAC[A/G]AAGGGGGACCACGAGATGGAGTCCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS45
PubMed Links
Gene Details
- Gene
- PRSS45
- Gene Name
- protease, serine 45
There are no transcripts associated with this gene.
- Gene
- PRSS46
- Gene Name
- protease, serine 46
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001205271.1 |
472 |
Silent Mutation |
TTC,TTT |
F143F |
NP_001192200.1 |
View Full Product Details