Product Details

SNP ID
rs192079267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70234061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTAGCTTCAGGGTTTTTTGTGTT[C/T]CCTTACCCATATCCATTTCGCCCAC
Phenotype
MIM: 601695 MIM: 607241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSN3 PubMed Links

Gene Details

Gene
CSN3
Gene Name
casein kappa
There are no transcripts associated with this gene.

Gene
FDCSP
Gene Name
follicular dendritic cell secreted protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152997.3 244 Silent Mutation TTC,TTT F44F NP_694542.1

View Full Product Details