Product Details
- SNP ID
-
rs192088020
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:71475588 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCTTTTCCCCTGTGTGAATACGC[C/T]GATGTTGGCTTAGATGGGAAGTCCT
- Phenotype
-
MIM: 194525
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF19
PubMed Links
Gene Details
- Gene
- ZNF19
- Gene Name
- zinc finger protein 19
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006961.3 |
1228 |
Missense Mutation |
CAG,CGG |
Q320R |
NP_008892.2 |
View Full Product Details