Product Details
- SNP ID
-
rs191637703
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:13153237 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCATCATGACCCTGTCGGTGTGGAA[A/G]ATGTATCACTCCCGCATGCAGCGCG
- Phenotype
-
MIM: 603765
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IER2
PubMed Links
Gene Details
- Gene
- IER2
- Gene Name
- immediate early response 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004907.2 |
379 |
Silent Mutation |
AAA,AAG |
K17K |
NP_004898.2 |
- Gene
- STX10
- Gene Name
- syntaxin 10
There are no transcripts associated with this gene.
View Full Product Details