Product Details

SNP ID
rs191676883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35924700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACTTCTAAAGCACAGAAGAACT[C/T]CAGGCTTTGCTTCGGAAGTGGCACC
Phenotype
MIM: 612751 MIM: 612896
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYZL6 PubMed Links

Gene Details

Gene
LYZL6
Gene Name
lysozyme like 6
There are no transcripts associated with this gene.

Gene
RDM1
Gene Name
RAD52 motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034836.1 521 Missense Mutation AAG,GAG K158E NP_001030008.1
NM_001163120.1 521 Intron NP_001156592.1
NM_001163121.1 521 Missense Mutation AAG,GAG K158E NP_001156593.1
NM_001163122.1 521 Missense Mutation AAG,GAG K135E NP_001156594.1
NM_001163124.1 521 Intron NP_001156596.1
NM_001163125.1 521 Intron NP_001156597.1
NM_001163130.1 521 Missense Mutation AAG,GAG K135E NP_001156602.1
NM_145654.3 521 Missense Mutation AAG,GAG K158E NP_663629.1
XM_011524509.2 521 Missense Mutation AAG,GAG K158E XP_011522811.1
XM_011524510.2 521 Intron XP_011522812.1

View Full Product Details