Product Details

SNP ID
rs192241917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:100646379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCTGCTGTTCTTCCTAACTCC[A/G]GCAGTGACACCAACATGGTATGCAG
Phenotype
MIM: 300642 MIM: 300191
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SRPX2 PubMed Links

Gene Details

Gene
SRPX2
Gene Name
sushi repeat containing protein, X-linked 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014467.2 537 Silent Mutation CCA,CCG P19P NP_055282.1
Gene
TSPAN6
Gene Name
tetraspanin 6
There are no transcripts associated with this gene.

View Full Product Details