Product Details

SNP ID
rs192624937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144130041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCACTGAGGAAGGAGAGGCTGA[C/T]GTGAGACATCTCAAAGGGAACCATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2A14 PubMed Links

Gene Details

Gene
OR2A14
Gene Name
olfactory receptor family 2 subfamily A member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001659.1 Intron NP_001001659.1

View Full Product Details