Product Details

SNP ID
rs192307797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:68789819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAAAAGGGTTATTGGTGCCATCG[C/G]GCAGGGTAGGTGAGCCCAGGAGGTC
Phenotype
MIM: 601064
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZFP36L1 PubMed Links

Gene Details

Gene
ZFP36L1
Gene Name
ZFP36 ring finger protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244698.1 1707 Missense Mutation CCC,CGC P244R NP_001231627.1
NM_001244701.1 1707 Missense Mutation CCC,CGC P313R NP_001231630.1
NM_004926.3 1707 Missense Mutation CCC,CGC P244R NP_004917.2

View Full Product Details